Z691372
Welch® peristaltic pump
analog, AC/DC input 115 V AC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
feature
CSA approved
manufacturer/tradename
Welch 3100B-01
parameter
0.003-0.3 L/min flow rate
20-200 rpm speed
Looking for similar products? Visit Product Comparison Guide
General description
Welch peristaltic pumps simplify your critical solution transfers. Avoid fluid cross-contamination while benefitting from precision delivery, handling material compatibility, and quick solution change-overs. Choose cartridges from four different tubing materials at one of three inner diameters for precision fluid deliveries ranging from 3 to 300 mL/min. Self-priming with easy adjustment of flow rate and pumping direction.
Starter Kit 3200K-01 is included with every pump
Initial Tubing Cartridges:
- Complete system - pump head, tubing & drive
- Lok-n-Flow tubing cartridges
- Bi-directional flow control & max flow button
- Variable speed
- Stackable & portable
Starter Kit 3200K-01 is included with every pump
Initial Tubing Cartridges:
- 1.6 mm silicone (2 each)
- 3.2 mm silicone (2 each)
- 4.8 mm silicone (2 each)
- 4.8 mm PVC Tubing (25 feet)
- Hose Clamps (4 each)
Legal Information
Welch is a registered trademark of Gardner Denver Thomas, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service