Documents
Z682357
Ace rotary evaporator condenser complete "V" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
plastic-coated glass
manufacturer/tradename
Büchi equivalent, 40603/32304
Related Categories
General description
This complete "V" evaporator condenser assembly includes:
The main glass condenser body with inner coil for vertical installation, O-ring, 1 L receiving flask, stainless steel clamp, GL threaded caps and hose barbs. Bottom joint is 35/20. It is designed to fit the Büchi® evaporator models 200/205 and series 114-144.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG