Key Documents
Z677957
Finntip® Flex pipette tips
Finntip Flex 300, non-sterile
Select a Size
960 EA
CHF 102.00
CHF 102.00
Estimated to ship on29 April 2025
Select a Size
Change View
960 EA
CHF 102.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
300 μL
volume range:
5-300 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 960 ea (Racks)
manufacturer/tradename:
Finn 94060510
CHF 102.00
Estimated to ship on29 April 2025
material
colorless tip
orange
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 ea (Racks)
manufacturer/tradename
Finn 94060510
maximum volume
300 μL
volume range
5-300 μL
General description
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)