Skip to Content
Merck
All Photos(2)

Documents

Z676764

ART® low retention barrier tips

Tip design ART 200, Hinged rack

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier
endotoxin free
Hinged rack
Tip design ART 200

packaging

pack of 4800 ea (50 hinged racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 2069-05-HR

technique(s)

PCR: suitable

maximum volume

200 μL

Looking for similar products? Visit Product Comparison Guide

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service