Documents
Z674370
Duran® GLS 80 caps and accessories
GL 18 pressure equalisation set for 4-port cap
Synonym(s):
1160167
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
manufacturer/tradename
DURAN® 1160167
General description
Flexible modular liquid or gas transfer 80 connection cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap.
Features and Benefits
• Manufactured from PP, PTFE, EPDM• The flexible modular system allows an individual combination of the different components according to requirements and application• Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be used by changing the inserts• Sterile pressure equalisation is possible by using the syringe filter• Unused ports can be sealed off with a blank GL 18 screw cap• Fully compatible with all the GLS 80 threads and therefore highly versatile
Legal Information
DURAN is a registered trademark of DWK Life Sciences