Key Documents
Z530255
Sigma-Aldrich
Aldrich® vacuum-transfer adapter
joint: ST/NS 14/20
Select a Size
1 EA
CHF 230.00
CHF 230.00
Estimated to ship on25 May 2025
Select a Size
Change View
1 EA
CHF 230.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
CHF 230.00
Estimated to ship on25 May 2025
Joint
joint: ST/NS14/20
stopcock size
2 mm
General description
PTFE stopcocks. For transfer of hazardous and sensitive reagents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)