Z511528
Kimax® heavy-duty wide-mouth, large numbers volumetric flasks - CLASS A
capacity 20 mL
Synonym(s):
92812G-20
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
glass
wide-mouth
Joint
joint ST/NS neck13
joint stopper13
sterility
non-sterile
feature
class A USP Type I and ASTM E438, Type I
closure type Glass stopper
packaging
pack of 1
case of 6
manufacturer/tradename
Kimble® 92812G-20
tol. measuring range
0.03 mL
capacity
20 mL
Looking for similar products? Visit Product Comparison Guide
General description
Wall thickness and weight are at least two times greater than that of standard volumetric flasks for durability. Calibrated "to contain". Meets ASTM E288, Class A, unserialized requirements. Easy pipette access.
Supplied with glass stopper.
Supplied with glass stopper.
Features and Benefits
• Manufactured from borosilicate glass 3.3• Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets• Heavy duty walls provide superior strength, durability and safety• Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills• Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets• Printed large volume numbers allow quick identification when selecting glassware• Flat interior bottoms for stirring with standard stir bars• Calibrate to contain• Graduated to Class A volumetric tolerances
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service