Key Documents
Z502642
Sigma-Aldrich
Threaded vial connector
8-425 thread for 8-425
Select a Size
1 EA
CHF 27.50
CHF 27.50
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 27.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
CHF 27.50
Check Cart for Availability
fitting
8-425 thread for 8-425
General description
These threaded connectors provide a convenient method to connect standard screw thread sample vials to glassware with GPI threads and to standard taper joint adapters.
• Excellent for collecting samples from rotary evaporator traps
• Made of glass filled nylon with inert PTFE inner seals
• Temp. range: up to 200 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)