Skip to Content
Merck
All Photos(1)

Documents

Z423483

Easy-Read® thermometer

temperature 20-440 °F, immersion level, total

Synonym(s):

immersion thermometer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112303
NACRES:
NB.79

feature

immersion level total

packaging

pack of 1 ea
pack of 25 ea

manufacturer/tradename

HB Instruments B60305-0100

temperature measuring range

20-440 °F

L

355 mm

O.D.

6-7 mm

Looking for similar products? Visit Product Comparison Guide

General description

Easy-to-read thermometers with black printing on a bright yellow background. Filled with contrasting black, non-hazardous, Enviro-Safe liquid. Calibrated with NIST, DKD standards.

Application

Easy-Read® thermometer can be used in pharmaceutical, petrochemical, food, beverage processing, dairies, breweries, distilleries, universities and chemical labs.

Legal Information

Easy Read is a registered trademark of SP Scienceware, a division of SP

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service