Key Documents
Z334510
Quickfit® three-neck round-bottom flasks with angled necks
center joint: ST/NS 14/23, side joint: ST/NS 14/23
Synonym(s):
FR25/1S/11A
Select a Size
1 EA
CHF 81.80
CHF 81.80
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 81.80
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
CHF 81.80
Check Cart for Availability
material
borosilicate glass
round bottom flask
three-neck
Joint
center joint: ST/NS14/23
side joint: ST/NS14/23
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Quickfit FR25/1S/11A
availability
not available in USA and Canada
H
77 mm
General description
With three necks fitted with Quickfit, precision-ground sockets. Axes of all three joints meet at bottom of flask.
  • One vertical neck
  • Two angled side necks
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)