Skip to Content
Merck
All Photos(1)

Key Documents

Z317756

Duran® vacuum desiccator replacement lids

knobbed, nominal diameter 150 mm , DN

Synonym(s):

244105706

Sign Into View Organizational & Contract Pricing

Select a Size

Z317756-1EA
CHF 128.00

CHF 128.00


Check Cart for Availability


Select a Size

Change View
Z317756-1EA
CHF 128.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

CHF 128.00


Check Cart for Availability

manufacturer/tradename

DURAN®
Schott

availability

not available in USA

nominal diameter

150 mm , DN

Looking for similar products? Visit Product Comparison Guide

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service