Z314366
FORTUNA® Optima glass syringes with interchangeable components
capacity 20 mL, glass Luer tip style
Select a Size
CHF 17.20
Select a Size
About This Item
CHF 17.20
Recommended Products
material
borosilicate glass
feature
autoclavable
graduated 1 mL
manufacturer/tradename
Fortuna Graf
capacity
20 mL
Related Categories
General description
- Interchangeable barrels and plungers
- Flanges flat on two sides to prevent syringe rolling
- Colored ring on plunger ends for easy volume measurement
- Conformity certified by H mark
Glass Luer - standard Luer hub
Luer lock - metal Luer lock tip on glass syringe, easy twist locks Luer needle onto syringe
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)