Z303445
Quickfit® two-neck, round-bottom flasks
capacity 250 mL, center joint: ST/NS 24/29
Synonym(s):
FR250/3S/2A
About This Item
Recommended Products
material
borosilicate glass
round bottom flask
two-neck (One vertical central neck and one angled side neck)
Joint
center joint: ST/NS24/29
side joint ST/NS19/26
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Quickfit FR250/3S/2A
H
130 mm
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service