Documents
Z268704
Scienceware® Safe-Lab PTFE stopper
joint: ST/NS 24/40, solid
Synonym(s):
lab stopper, lab stoppers, stoppers
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
Joint
joint: ST/NS24/40
packaging
pkg of 2 ea
manufacturer/tradename
Bel-Art F20196-1502
General description
PTFE stoppers with PP extracting nut for easy removal from flask. For use with boiling and distilling flasks with ground joints.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP