Z261025
Nalgene® polypropylene copolymer separatory funnel
capacity 250 mL
Synonym(s):
liquid separation funnel, plastic separation device
All Photos(1)
About This Item
Recommended Products
material
polypropylene copolymer bottle (PPCO)
packaging
pack of 1 ea
manufacturer/tradename
Nalgene 4300-0250
capacity
250 mL
closure size
33 mm
Related Categories
General description
Break-resistant and nearly transparent. Phase interface visible down to stopcock. PTFE stopcock and Tefzel® screw closure are leakproof.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company