Key Documents
Z256021
Gum rubber laboratory tubing
tubing I.D. × O.D. 3/4 in. × 1 1/2 in.
Select a Size
10 FT
CHF 223.00
CHF 223.00
Check Cart for Availability
Select a Size
Change View
10 FT
CHF 223.00
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
CHF 223.00
Check Cart for Availability
description
heavy wall
packaging
pkg of 10 feet
tubing I.D. × O.D.
3/4 in. × 1 1/2 in.
wall size
3/8 in.
General description
Tubing is red, and can be used for pressure or vacuum work.
Choose from one of the most recent versions:
Certificates of Analysis (COA)