Key Documents
Z228117
Coverspec eyeshields
polycarbonate
About This Item
UNSPSC Code:
46181802
material
polycarbonate
manufacturer/tradename
Bollé
availability
not available in USA and Canada
General description
Ideal for laboratory visitors and occasional users, this ultra-lightweight eyeshield with a single piece polycarbonate lens can be worn over most prescripton spectacles. BS2092.2
Choose from one of the most recent versions:
Certificates of Analysis (COA)