Key Documents
Z225568
Engraving pen
carbide tip, pkg of 1 ea
Select a Size
1 EA
CHF 36.10
CHF 36.10
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 36.10
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
CHF 36.10
Check Cart for Availability
packaging
pkg of 1 ea
General description
Carbide-tip pen with metal body and clip.
Application
Engraving pen has been used:
  • to label slides for DNA combing
  • to cut the coverslips into small pieces for microscopy imaging
  • to encircle cryopreserved sections on glass slides
Choose from one of the most recent versions:
Certificates of Analysis (COA)