Documents
Z186821
Sigma-Aldrich
All-purpose sampling containers
capacity 4 1/2 oz
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
feature
lid: no
capacity
4 1/2 oz
General description
Durable and translucent PP containers resist acids, bases, most organic solvents, and common laboratory reagents. Internally molded dual graduations in increments of 1 oz and 20 mL. Economical enough to discard after one use, or strong enough to reuse. Tight-fitting PE lid not included. Nonsterile.
Other Notes
Supplied without lid. No lid available.