Skip to Content
Merck
All Photos(1)

Documents

Z183377

Foil/poly bag

size 2 in. × 4 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24111500
NACRES:
NB.35

packaging

pkg of 75 ea

size

2 in. × 4 in.

thickness

5 mil

Looking for similar products? Visit Product Comparison Guide

General description

Heat sealable. Useful for storing air, light, or moisture-sensitive reagents, odoriferous products, and for temporarily containing leaking bottles.

Recommended heat seal conditions: 400°F (204°C), 40 psi, 1.5 sec.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service