Documents
Z171697
Ace double-jacketed coiled condenser
jacket L 300 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 595514
jacket L
300 mm
Related Categories
General description
Extra cooling capacity for volatile materials. Compact design retards diffusion.