Key Documents
Z152447
Sigma-Aldrich
Aldrich® dry ice condenser for Büchi® rotary evaporators
with feed tube
Select a Size
1 EA
CHF 1’120.00
CHF 1’120.00
Check Cart for Availability
Select a Size
Change View
1 EA
CHF 1’120.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
CHF 1’120.00
Check Cart for Availability
Joint
male joint SJ35/20
joint stopcock19/38
volume measuring range
500 mL (dry ice reservoir)
General description
For Büchi rotary evaporators R110, and R151. For use with short stem duct.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)