Z108898
Mini Diazald® apparatus
joint: ST/NS 19/22 Clear-Seal joints, for generating 1 to 50 mmol of diazomethane
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS19/22 Clear-Seal joints
Related Categories
General description
For technical and safety information about Diazald and Aldrich diazomethane generators see Technical Information Bulletin AL-180.
The mini Diazald apparatus bridges the gap between the System 45 diazomethane generator Z411736 (for preparing <1 mmol of diazomethane) and the Diazald kit (for preparing ~100 mmol of diazomethane). It consists of a reaction vessel and a condenser in one compact unit. This apparatus features a cold-finger in place of a water-jacketed condenser. When filled with dry ice/acetone slush, the condenser efficiently prevents dangerous diazomethane/ether vapor from escaping. A separatory funnel, Z100382 and a round-bottom receiving flask, Z100358, (must be equipped with Clear-Seal joints) are the only other glassware needed and can be purchased separately.
NOTE: Sigma-Aldrich is unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
NOTE: Sigma-Aldrich is unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
Legal Information
Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany
required but not provided
Customers Also Viewed
Journal of medicinal chemistry, 59(17), 7856-7876 (2016-07-28)
Targeted antifolates with heteroatom replacements of the carbon vicinal to the phenyl ring in 1 by N (4), O (8), or S (9), or with N-substituted formyl (5), acetyl (6), or trifluoroacetyl (7) moieties, were synthesized and tested for selective