WHA1950207
Whatman® glass microfiber filtration funnel
capacity 115 mL , reservoir, diam. 70 mm
Synonym(s):
Z753238
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pk of 1
manufacturer/tradename
Whatman 1950-207
Whatman Article No. 28418634 (US reference)
capacity
115 mL , reservoir
115 mL
diam.
70 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service