Key Documents
WHA10461100
Whatman® FP 050/0 Filter holder
Synonym(s):
Z698512
About This Item
UNSPSC Code:
41104922
NACRES:
NB.24
Pricing and availability is not currently available.
packaging
pkg of 1 ea
manufacturer/tradename
Whatman 10461100
Whatman Article No. 28415719 (US reference)
General description
The female Luer lock inlet is compatible with typical syringes. Polysulfone housing can be autoclaved with membrane in place for applications requiring a sterile filter assembly.
Features and Benefits
  • Durable polysulfone plastic construction provides moderate chemical compatibility with lighter weight than stainless steel filter holders.
  • Polysulfone filter holders are often used for clarification, venting applications, particulate removal or collection from aqueous, air or gas streams.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)