Skip to Content
Merck
All Photos(1)

Documents

WHA10382514

Whatman® cellulose chromatography papers

2727 circles, diam. 185 mm

Synonym(s):

Z750166

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115727
NACRES:
NB.24

material

cellulose membrane
circles

grade

for chromatography

sterility

non-sterile

feature

2727-CHR

packaging

pk of 100

manufacturer/tradename

Whatman 10382514
Whatman Article No. 28414860 (US reference)

parameter

3.6 cm/hr speed
fast flow rate

diam.

185 mm

basis weight

700 g/m2

Looking for similar products? Visit Product Comparison Guide

General description

2727 Chr cellulose chromatography sheets is a 1.40 mm paper for separation of very large amounts of substance. Thickness 1.40 mm. Flow rate 180 mm/30 min. For separation of very large amounts of substance.

Features and Benefits

  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Campuzano et al.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service