W524107
3-[5,5,6-Trimethylbicyclo[2.2.1]hept-2-yl]cyclohexan-1-ol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C16H28O
CAS Number:
Molecular Weight:
236.39
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
biological source
synthetic
refractive index
n20/D 1.496 (lit.)
bp
302 °C (lit.)
density
0.97 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Organoleptic
ethereal
SMILES string
CC1[C@@H]2C[C@@H](CC2C3CCCC(O)C3)C1(C)C
InChI
1S/C16H28O/c1-10-14-8-12(16(10,2)3)9-15(14)11-5-4-6-13(17)7-11/h10-15,17H,4-9H2,1-3H3/t10?,11?,12-,13?,14-,15?/m0/s1
InChI key
BWVZAZPLUTUBKD-HXLKAFCPSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service