Skip to Content
Merck
All Photos(1)

Key Documents

W425301

Sigma-Aldrich

Methyl 10-undecenoate

96%

Synonym(s):

10-Undecenoic acid methyl ester, Undecylenic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)8CO2CH3
CAS Number:
Molecular Weight:
198.30
FEMA Number:
4253
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

reg. compliance

FDA 21 CFR 117

Assay

96%

refractive index

n20/D 1.437 (lit.)

bp

245 °C (lit.)

solubility

H2O: insoluble
alcohol: soluble
oil: soluble
propylene glycol: soluble

density

0.882 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; fruity; waxy

SMILES string

COC(=O)CCCCCCCCC=C

InChI

1S/C12H22O2/c1-3-4-5-6-7-8-9-10-11-12(13)14-2/h3H,1,4-11H2,2H3

InChI key

KISVAASFGZJBCY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service