Skip to Content
Merck
All Photos(1)

Key Documents

W424701

Sigma-Aldrich

Tripropylamine

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)3N
CAS Number:
Molecular Weight:
143.27
FEMA Number:
4247
Beilstein:
1098331
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
11.026

biological source

synthetic

vapor density

4.9 (vs air)

vapor pressure

2.9 mmHg ( 20 °C)

Assay

≥98%

autoignition temp.

356 °F

expl. lim.

5.6 %

refractive index

n20/D 1.416 (lit.)

bp

155-158 °C (lit.)
40-42 °C/11 mmHg (lit.)

mp

−93.5 °C (lit.)

density

0.753 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

fishy

SMILES string

CCCN(CCC)CCC

InChI

1S/C9H21N/c1-4-7-10(8-5-2)9-6-3/h4-9H2,1-3H3

InChI key

YFTHZRPMJXBUME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueshu Li et al.
Langmuir : the ACS journal of surfaces and colloids, 28(9), 4152-4159 (2012-02-07)
The micelles of two tripropargylammonium-functionalized cationic surfactants were cross-linked by a disulfide-containing diazido cross-linker in the presence of Cu(I) catalysts. With multiple residual alkyne groups on the surface, the resulting surface cross-linked micelles (SCMs) were postfunctionalized by reaction with 2-azidoethanol
H C Froede et al.
Archives of biochemistry and biophysics, 247(2), 420-423 (1986-06-01)
The theory of noncompetitive inhibition of acetylcholinesterase based on the binding of inhibitor to the acetylenzyme and the free enzyme was proven correct by demonstrating that tripropylammonium ion increases the steady-state concentration of acetylenzyme, as predicted by the theory. By
Xiang Huang et al.
The Analyst, 140(17), 6015-6024 (2015-07-21)
An efficient aptasensor was developed in which graphene oxide (GO) was employed as an indicator for both electrochemical impedance spectroscopy and electrochemiluminescence (ECL) signal generation. The aptasensor was fabricated by self-assembling the ECL probe of a thiolated adenosine triphosphate binding
Anna-Maria Spehar-Délèze et al.
Analytical and bioanalytical chemistry, 407(22), 6657-6667 (2015-06-24)
Development of a fully automated electrochemiluminescence (ECL) DNA assay for multiplex detection of six biowarfare agents is described. Aminated-DNA capture probes were covalently immobilised on activated-carbon electrodes and subsequently hybridised to target strands. Detection was achieved via a sandwich-type assay
Limin Zhou et al.
ACS applied materials & interfaces, 7(44), 24438-24445 (2015-10-22)
Ru(bpy)3(2+)-doped silica (Ru-SiO2) nanoparticles and gold-nanoparticle-decorated graphene (P-RGO@Au) were combined to form a P-RGO@Au@Ru-SiO2 composite. The composite was used to develop a novel sandwich-type electrochemiluminescence immunosensor for the analysis of HIV-1 p24 antigen. The composite worked as carrier to immobilize

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service