Skip to Content
Merck
All Photos(1)

Key Documents

W379603

Sigma-Aldrich

Vanillyl butyl ether

≥96%

Synonym(s):

4-(butoxymethyl)-2-methoxy-Phenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H3-3-(OCH3)CH2O(CH2)3CH3
CAS Number:
Molecular Weight:
210.27
FEMA Number:
3796
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.093
Organoleptic:
creamy
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

Assay

≥96%

refractive index

n20/D 1.516 (lit.)

bp

241 °C (lit.)

density

1.057 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy

SMILES string

CCCCOCC1=CC(OC)=C(O)C=C1

InChI

1S/C12H18O3/c1-3-4-7-15-9-10-5-6-11(13)12(8-10)14-2/h5-6,8,13H,3-4,7,9H2,1-2H3

InChI key

VLDFMKOUUQYFGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vanillyl butyl ether is an alkoxy-substituted benzyl derivative mainly used as a flavoring agent.[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam TB, et al.
Adam TB, et al.
Food And Chemical Toxicology, 43(8), 1241-1271 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service