Skip to Content
Merck
All Photos(1)

Documents

W369918

Sigma-Aldrich

Isopropyl 2-methylbutyrate

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2CH(CH3)2
CAS Number:
Molecular Weight:
144.21
FEMA Number:
3699
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.547
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Kosher

Agency

meets purity specifications of JECFA

Assay

≥98%

refractive index

n20/D 1.397 (lit.)

bp

140-144 °C/727 mmHg (lit.)

density

0.851 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; oily; fruity; ethereal; sweet

SMILES string

CCC(C)C(=O)OC(C)C

InChI

1S/C8H16O2/c1-5-7(4)8(9)10-6(2)3/h6-7H,5H2,1-4H3

InChI key

DIRDKDDFAMNBNY-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service