Skip to Content
Merck
All Photos(1)

Key Documents

W358315

Sigma-Aldrich

3-Octyl acetate

natural (US), ≥97%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2CH(C2H5)(CH2)4CH3
CAS Number:
Molecular Weight:
172.26
FEMA Number:
3583
EC Number:
Council of Europe no.:
2347
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.254
grade:
FG
Halal
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 173.65 (172.515)

Assay

≥97%

refractive index

n20/D 1.415 (lit.)

bp

99 °C/18 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; green; citrus; waxy; minty

SMILES string

CCCCCC(CC)OC(C)=O

InChI

1S/C10H20O2/c1-4-6-7-8-10(5-2)12-9(3)11/h10H,4-8H2,1-3H3

InChI key

STZUZYMKSMSTOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service