Skip to Content
Merck
All Photos(1)

Documents

W354708

Sigma-Aldrich

3-Heptanol

≥98%

Synonym(s):

Butyl ethyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH2CH3
CAS Number:
Molecular Weight:
116.20
FEMA Number:
3547
Beilstein:
1719067
EC Number:
Council of Europe no.:
544
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.044
NACRES:
NA.21

biological source

synthetic

Assay

≥98%

refractive index

n20/D 1.421 (lit.)

bp

66 °C/20 mmHg (lit.)

density

0.821 g/mL at 20 °C
0.818 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous

SMILES string

CCCCC(O)CC

InChI

1S/C7H16O/c1-3-5-6-7(8)4-2/h7-8H,3-6H2,1-2H3

InChI key

RZKSECIXORKHQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Odor at 1.0%
Taste at 2 ppm

Other Notes

Natural occurrence: Banana cranberry papaya potato, malt, peppermint, Scotch spearmint, butter, cooked beef, coffee and brandy.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mosciano, G.
Perfumer & Flavorist, 27(4), 68-68 (2002)
Maria Stecklum et al.
In vitro cellular & developmental biology. Animal, 51(2), 183-191 (2014-10-02)
In the present study, purified human cord blood stem cells were co-cultivated with murine hepatic alpha mouse liver 12 (AML12) cells to compare the effect on endodermal stem cell differentiation by either direct cell-cell interaction or by soluble factors in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service