Skip to Content
Merck
All Photos(1)

Key Documents

W349700

Sigma-Aldrich

cis-3-Hexenyl 2-methylbutanoate

FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3497
EC Number:
Council of Europe no.:
2345
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.854
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

Assay

≥97%

refractive index

n20/D 1.432 (lit.)

density

0.878 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; fresh; green; fruity; pear; sweet

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)C(C)CC

InChI

1S/C11H20O2/c1-4-6-7-8-9-13-11(12)10(3)5-2/h6-7,10H,4-5,8-9H2,1-3H3/b7-6-

InChI key

JKKGTSUICJWEKB-SREVYHEPSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service