Skip to Content
Merck
All Photos(1)

Key Documents

W348902

Sigma-Aldrich

Ethyl 2-methyl-4-pentenoate

≥98%, stabilized, FG

Synonym(s):

pineapple pentenoate

Sign Into View Organizational & Contract Pricing

Select a Size

W348902-SAMPLE-K
CHF 52.50
100 G
CHF 186.00

CHF 52.50


Estimated to ship on23 March 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W348902-SAMPLE-K
CHF 52.50
100 G
CHF 186.00

About This Item

Linear Formula:
H2C=CHCH2CH(CH3)CO2C2H5
CAS Number:
Molecular Weight:
142.20
FEMA Number:
3489
EC Number:
Council of Europe no.:
10613
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.527
NACRES:
NA.21
Organoleptic:
green; tropical; fruity; cooling; pineapple
grade:
FG
Halal
Kosher
biological source:
synthetic
Agency:
meets purity specifications of JECFA
food allergen:
no known allergens

CHF 52.50


Estimated to ship on23 March 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

contains

α-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.417 (lit.)

bp

153-155 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; tropical; fruity; cooling; pineapple

SMILES string

CCOC(=O)C(C)CC=C

InChI

1S/C8H14O2/c1-4-6-7(3)8(9)10-5-2/h4,7H,1,5-6H2,2-3H3

InChI key

BDBGKYIBDXAVMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service