Skip to Content
Merck
All Photos(1)

Key Documents

W348406

Sigma-Aldrich

1,2-Ethanedithiol

≥98%

Synonym(s):

1,2-Dimercaptoethane, Dithioglycol, Ethylene mercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2SH
CAS Number:
Molecular Weight:
94.20
FEMA Number:
3484
Beilstein:
505946
EC Number:
Council of Europe no.:
11467
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.066

grade

Kosher

vapor density

>1 (vs air)

vapor pressure

4.8 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.558 (lit.)

bp

144-146 °C (lit.)

mp

−41 °C (lit.)

density

1.123 g/mL at 25 °C (lit.)

SMILES string

SCCS

InChI

1S/C2H6S2/c3-1-2-4/h3-4H,1-2H2

InChI key

VYMPLPIFKRHAAC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cândida F Pereira et al.
PloS one, 6(2), e17016-e17016 (2011-02-25)
Due to its small size and versatility, the biarsenical-tetracysteine system is an attractive way to label viral proteins for live cell imaging. This study describes the genetic labeling of the human immunodeficiency virus type 1 (HIV-1) structural proteins (matrix, capsid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service