Skip to Content
Merck
All Photos(1)

Key Documents

W343005

Sigma-Aldrich

4-Hexen-1-ol

≥96%, predominantly trans, FG

Synonym(s):

4-Hexen-1-ol, predominantly trans

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CH(CH2)3OH
CAS Number:
Molecular Weight:
100.16
FEMA Number:
3430
EC Number:
Council of Europe no.:
2295
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.074
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥96%

refractive index

n20/D 1.439 (lit.)

bp

159-160 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; musty; herbaceous; tomato

SMILES string

C\C=C\CCCO

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h2-3,7H,4-6H2,1H3/b3-2+

InChI key

VTIODUHBZHNXFP-NSCUHMNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Hexen-1-ol is a volatile flavor alcohol that is reported to occur in banana.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penicillin acylase-mediated synthesis of 2-acetyl-1-pyrroline and of 2-propionyl-1-pyrroline, key roast-smelling odorants in food. Inclusion complexes with ?-cyclodextrin and their NMR and MS characterization.
Favino TF, et al.
The Journal of Organic Chemistry, 61(25), 8975-8979 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service