W337404
3-(Methylthio)butanal
≥96%, FG
Synonym(s):
3-methylsulfanylbutanal, Potato butyraldehyde
About This Item
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Halal
Kosher
Agency
meets purity specifications of JECFA
reg. compliance
EU Regulation 1334/2008 & 178/2002
vapor density
4.1 (vs air)
vapor pressure
60 mmHg ( 20 °C)
description
may contain approx. 3% Crotonaldehyde
Assay
≥96%
impurities
<4% 2-butenal
refractive index
n20/D 1.476 (lit.)
bp
62-64 °C/10 mmHg (lit.)
density
1.001 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
green; vegetable; potato; sulfurous
storage temp.
2-8°C
SMILES string
CSC(C)CC=O
InChI
1S/C5H10OS/c1-5(7-2)3-4-6/h4-5H,3H2,1-2H3
InChI key
NCBDFIPMWRKPDU-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service