Skip to Content
Merck
All Photos(1)

Key Documents

W335401

Sigma-Aldrich

Hexyl trans-2-butenoate

≥95%, FCC

Synonym(s):

Hexyl crotonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCO2(CH2)5CH3
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3354
EC Number:
Council of Europe no.:
10688
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.266

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FCC

Assay

≥95%

refractive index

n20/D 1.438 (lit.)

density

0.885 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

pineapple

SMILES string

[H]\C(C)=C(\[H])C(=O)OCCCCCC

InChI

1S/C10H18O2/c1-3-5-6-7-9-12-10(11)8-4-2/h4,8H,3,5-7,9H2,1-2H3/b8-4+

InChI key

MZNHUHNWGVUEAT-XBXARRHUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

192.2 °F

Flash Point(C)

89 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service