Skip to Content
Merck
All Photos(1)

Key Documents

W333200

Sigma-Aldrich

Butan-3-one-2-yl butanoate

≥98%, FCC, FG

Synonym(s):

acetoin butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH(CH3)COCH3
CAS Number:
Molecular Weight:
158.19
FEMA Number:
3332
EC Number:
Council of Europe no.:
10525
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.264
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥98%

refractive index

n20/D 1.4185 (lit.)

density

0.982 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; musty; buttery

SMILES string

CCCC(=O)OC(C)C(C)=O

InChI

1S/C8H14O3/c1-4-5-8(10)11-7(3)6(2)9/h7H,4-5H2,1-3H3

InChI key

LJDWJXUIGKSETE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.6 °F

Flash Point(C)

80.9 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service