Skip to Content
Merck
All Photos(1)

Documents

W324110

Sigma-Aldrich

Trimethylamine solution

45 wt. % in H2O, FG

Synonym(s):

N,N-dimethylmethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
FEMA Number:
3241
Beilstein:
956566
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Flavis number:
11.009
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002

concentration

45 wt. % in H2O

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy; meaty; oily

SMILES string

CN(C)C

InChI

1S/C3H9N/c1-4(2)3/h1-3H3

InChI key

GETQZCLCWQTVFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-49.0 °F - closed cup

Flash Point(C)

-45 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service