Skip to Content
Merck
All Photos(1)

Key Documents

W311308

Sigma-Aldrich

Wintergreen oil

China origin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
3113
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

Gaultheria procumbens L.

grade

Kosher

optical activity

[α]20/D −0.5°, neat

origin

China origin

bp

219 °C (lit.)

density

1.181 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; sweet; wintergreen

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: steam distillation

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ticiano P Barbosa et al.
Bioorganic & medicinal chemistry, 19(14), 4250-4256 (2011-06-21)
The chalcone-like series 1a-1g was efficiently synthesized from Morita-Baylis-Hillman reaction (52-74% yields). Compounds 1a-1g were designed by molecular hybridization based on the anti-inflammatory drug methyl salicylate (3) and the antileishmanial moiety of the Morita-Baylis-Hillman adducts 2a-2g. The 1a-1g compounds were
Michael Jones et al.
Journal of medicinal chemistry, 52(21), 6588-6598 (2009-10-14)
Aspirin prodrugs and related nitric oxide releasing compounds hold significant therapeutic promise, but they are hard to design because aspirin esterification renders its acetate group very susceptible to plasma esterase mediated hydrolysis. Isosorbide-2-aspirinate-5-salicylate is a true aspirin prodrug in human
Jihye Kim et al.
European journal of medicinal chemistry, 48, 36-44 (2011-12-14)
To develop a more potent NFκB inhibitor from salicylic acid which is known to inhibit activity of NFκB, a transcription factor regulating genes involved in immunity, inflammation and tumorigenesis, derivatives of salicylic acid (SA) where the 5 position, carboxyl or
Lourdes Santana et al.
Journal of medicinal chemistry, 51(21), 6740-6751 (2008-10-07)
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives
Denis Fourches et al.
Chemical research in toxicology, 23(1), 171-183 (2009-12-18)
Drug-induced liver injury is one of the main causes of drug attrition. The ability to predict the liver effects of drug candidates from their chemical structures is critical to help guide experimental drug discovery projects toward safer medicines. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service