Skip to Content
Merck
All Photos(1)

Key Documents

W287008

Sigma-Aldrich

Phenethyl tiglate

≥95%, stabilized, FG

Synonym(s):

Benzyl carbinyl tiglate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH3)CO2CH2CH2C6H5
CAS Number:
Molecular Weight:
204.26
FEMA Number:
2870
EC Number:
Council of Europe no.:
2186
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.496
NACRES:
NA.21

Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥95%

contains

α-tocopherol as stabilizer (synthetic)

refractive index

n20/D 1.514 (lit.)

bp

139-140 °C/7 mmHg (lit.)

density

1.018 g/mL at 25 °C (lit.)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sapir Penker et al.
Frontiers in systems neuroscience, 14, 604718-604718 (2020-12-18)
Coding of odorous stimuli has been mostly studied using single isolated stimuli. However, a single sniff of air in a natural environment is likely to introduce airborne chemicals emitted by multiple objects into the nose. The olfactory system is therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service