Skip to Content
Merck
All Photos(1)

Key Documents

W238708

Sigma-Aldrich

2′,4′-Dimethylacetophenone

≥96%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3COCH3
CAS Number:
Molecular Weight:
148.20
FEMA Number:
2387
Beilstein:
956772
EC Number:
Council of Europe no.:
157
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.023

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥96%

refractive index

n20/D 1.535 (lit.)

bp

117-118 °C/18 mmHg (lit.)

density

0.997 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

floral; woody; sweet

SMILES string

CC(=O)c1ccc(C)cc1C

InChI

1S/C10H12O/c1-7-4-5-10(9(3)11)8(2)6-7/h4-6H,1-3H3

InChI key

HSDSKVWQTONQBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2′,4′-Dimethylacetophenone can be used as a flavoring agent. It is reported to be found in the volatiles of Koshu wine.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock, GA
Encyclopedia of Food and Color Additives, 1, 844-845 (1997)
Burdock, GA
Encyclopedia of Food and Color Additives, 1, 844-845 (1997)
Volatile components identified in the phenolic fractions of wines from Koshu and Zenkoji grapes.
Shimizu JI & Watanabe M
Agricultural and Biological Chemistry, 46(6), 1447-1452 (1982)
Masahiko Nakamura et al.
Bioorganic & medicinal chemistry, 17(13), 4740-4746 (2009-05-16)
Adult T cell leukemia (ATL), caused by infection of human T-lymphotropic virus type 1 (HTLV-1), has a poor prognosis and curative therapy is unavailable, so it is important to find or design superior lead compounds for the drug treatment of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service