Skip to Content
Merck
All Photos(1)

Key Documents

W220213

Sigma-Aldrich

Isobutyl hexanoate

natural, ≥95%

Synonym(s):

Isobutyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2202
EC Number:
Council of Europe no.:
314c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.064

grade

Halal
Kosher
natural

reg. compliance

FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.414 (lit.)

bp

348-350 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple

SMILES string

CCCCCC(=O)OCC(C)C

InChI

1S/C10H20O2/c1-4-5-6-7-10(11)12-8-9(2)3/h9H,4-8H2,1-3H3

InChI key

UXUPPWPIGVTVQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service