All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H14O3
CAS Number:
Molecular Weight:
182.22
FEMA Number:
2072
EC Number:
Council of Europe no.:
2109
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.025
biological source
synthetic
Assay
≥98%
refractive index
n20/D 1.474 (lit.)
bp
95-97 °C/1 mmHg (lit.)
density
1.033 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Organoleptic
fatty; oily
SMILES string
CCCCCOC(=O)c1ccco1
InChI
1S/C10H14O3/c1-2-3-4-7-13-10(11)9-6-5-8-12-9/h5-6,8H,2-4,7H2,1H3
InChI key
BGJZMKPSRLHMME-UHFFFAOYSA-N
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service