TEM-LC200-NI-150
Lacey Carbon Supported Nickel Grid
size 200 mesh, hole size 150 μm , Average, box of 25
All Photos(1)
About This Item
UNSPSC Code:
41111761
Recommended Products
packaging
box of 25
hole size
150 μm , Average
size
200 mesh
General description
Lacey Carbon Supported Grid: Nickel Grid, 200 mesh. Average hole size is 150 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)