Key Documents
TEM-CF400NI50
Sigma-Aldrich
Carbon Film Supported Nickel Square Mesh
size 400 mesh, box of 50
Synonym(s):
CARBON FILM 400 MESH NI 50/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-CF400NI50 is currently not available for purchase. Contact Technical Service
packaging
box of 50
size
400 mesh
General description
A 5-6 nm thin film of pure carbon supported standard squre mesh grids. Nickel grid, 400 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)