Key Documents
TEM-CF200F2-CU-ET
Sigma-Aldrich
Carbon Film Supported Copper Gilder Finder F2 Grids
Extra Thick, size 200 mesh, box of 50
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
200 mesh
General description
A 20-30 nm thick film of pure carbon supported glider finder (F2) grids. Copper grid, 200 mesh. Gilder finder grids are locator grids with reference/index on the grid: 3.05mm diameter, 0.7 mil (18μm) thickness. F2 type of design has thick bars dividing regions into 9 thin bar areas, which are identified by alphabetical letters located in the center of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)