Skip to Content
Merck
All Photos(1)

Key Documents

SYNV415045

Synthware vacuum trap complete

joint: ST/NS 35/20

Synonym(s):

Synthware V415045

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 177.00

CHF 177.00


Check Cart for Availability


Select a Size

Change View
1 EA
CHF 177.00

About This Item

UNSPSC Code:
40161501
NACRES:
NB.51

CHF 177.00


Check Cart for Availability

Joint

joint: ST/NS35/20

manufacturer/tradename

V415045

General description

For use with vacuum manifolds having 35/20 socket joints. Please order tubing adapters and ball joint clamp separately.

Accessories:
SYNV175037 Top tubing adapter
SYNV175038 Middle tubing adapter
SYNV175039 Bottom tubing adapter
Z154075 Clamp for 35/20 ball joint

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service